5-Chloro-7-azaindole, 95%, Thermo Scientific Chemicals
5-Chloro-7-azaindole, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Chloro-7-azaindole, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66268.03
also known as H66268-03
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS866546-07-8
IUPAC Name5-chloro-1H-pyrrolo[2,3-b]pyridine
Molecular FormulaC7H5ClN2
InChI KeyMFZQJIKENSPRSJ-UHFFFAOYSA-N
SMILESClC1=CN=C2NC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Appearance (Color)Pale yellow.
Assay (unspecified)Water: ≤0.5%
Assay from Supplier's CofA≥94.0%
Melting Point162.0-166.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only