2-(Bromomethyl)oxetane, 96%, Thermo Scientific Chemicals
2-(Bromomethyl)oxetane, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Bromomethyl)oxetane, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H66276.03
also known as H66276-03
Price (USD)/ Each
954.65
Online exclusive
1060.00 
Save 105.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
954.65
Online exclusive
1060.00 
Save 105.35 (10%)
Add to cart
2-(Bromomethyl)oxetane, 96%, Thermo Scientific Chemicals
Catalog numberH66276.03
Price (USD)/ Each
954.65
Online exclusive
1060.00 
Save 105.35 (10%)
-
Add to cart
Chemical Identifiers
CAS939759-23-6
IUPAC Name2-(bromomethyl)oxetane
Molecular FormulaC4H7BrO
InChI KeyOMXAGUVERXNCSZ-UHFFFAOYNA-N
SMILESBrCC1CCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to pale yellow.
FormLiquid.
Assay (GC)≥95.0%
Proton NMRConforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only