Benzyl 3-bromopropyl ether may be used in the studies of preparation of (2S,3S)-1-[(triisopropylsilyl)oxy]-7-(benzyloxy)-2,3-(isopropylidenedioxy)-4(Z)-heptene, Ppeparation of 5-(3-Benzyloxypropoxy)psoralen (PAP-7), total synthesis of zincophorin and (+)-anatoxin-a.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzyl 3-bromopropyl ether may be used in the studies of preparation of (2S,3S)-1-[(triisopropylsilyl)oxy]-7-(benzyloxy)-2,3-(isopropylidenedioxy)-4(Z)-heptene, Ppeparation of 5-(3-Benzyloxypropoxy)psoralen (PAP-7), total synthesis of zincophorin and (+)-anatoxin-a.
Solubility
Slightly soluble in water.
Notes
Store under dry inert gas. Store away from air. Incompatible with air and oxidizing agents.
RUO – Research Use Only
Jack Taunton; Jon L. Collins and Stuart L. Schreiber. Synthesis of Natural and Modified Trapoxins, Useful Reagents for Exploring Histone Deacetylase Function. J. Am. Chem. Soc.1996, 116, (43), 10412-10422.
Magali Defosseux; Nicolas Blanchard; Christophe Meyer; Janine Cossy. Total synthesis of zincophorin and its methyl ester. Journal of Organic Chemistry.2004, 69, (14), 4626-4647.