Thermo Scientific Chemicals

7-(Fmoc-amino)heptanoic acid, 95%, Thermo Scientific Chemicals

Catalog number: H66285.03
1 g, Each
Thermo Scientific Chemicals

7-(Fmoc-amino)heptanoic acid, 95%, Thermo Scientific Chemicals

Catalog number: H66285.03
1 g, Each
Quantity
Catalog number: H66285.03
also known as H66285-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
127582-76-7
IUPAC Name
7-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)heptanoic acid
Molecular Formula
C22H25NO4
InChI Key
FRPYXTFBPCYALT-UHFFFAOYSA-N
SMILES
OC(=O)CCCCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Assay (HPLC)
>94.0%

Description

7-(Fmoc-amino)heptanoic acid is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-(Fmoc-amino)heptanoic acid is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Store at 4°C. Incompatible with heat and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text