N-Boc-DL-methionine, 98%, Thermo Scientific Chemicals
N-Boc-DL-methionine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-DL-methionine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H66372.06
also known as H66372-06
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS93000-03-4
IUPAC Name2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
Molecular FormulaC10H19NO4S
InChI KeyIMUSLIHRIYOHEV-UHFFFAOYNA-N
SMILESCSCCC(NC(=O)OC(C)(C)C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (HPLC)≥97.5%
Solution TestClear solution (0.3 g in 2mL DMF)
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only