Thermo Scientific Chemicals

Fluorolink|r E10-H, Thermo Scientific Chemicals

Catalog number: H66374.14
25 g, Each
Thermo Scientific Chemicals

Fluorolink|r E10-H, Thermo Scientific Chemicals

Catalog number: H66374.14
25 g, Each
Quantity
Catalog number: H66374.14
also known as H66374-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
162492-15-1
Color (APHA)
≤100
Appearance (Color)
Clear
Form
Liquid
Water Content
≤500 ppm
Equivalent Weight
800-1100 AMW

Description

Use in polymer modifications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Use in polymer modifications.

Solubility
Soluble in fluorinated solvents, alcohol, and ketones. Insoluble in water.

Notes
Incompatible with heat and oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text