N-[3-(Trimethoxysilyl)propyl]-N,N,N-trimethylammonium chloride, 50% in methanol, Thermo Scientific Chemicals
N-[3-(Trimethoxysilyl)propyl]-N,N,N-trimethylammonium chloride, 50% in methanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-[3-(Trimethoxysilyl)propyl]-N,N,N-trimethylammonium chloride, 50% in methanol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number H66414.22
also known as H66414-22
Price (USD)/ Each
72.65
Online exclusive
80.00 
Save 7.35 (9%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
72.65
Online exclusive
80.00 
Save 7.35 (9%)
Add to cart
N-[3-(Trimethoxysilyl)propyl]-N,N,N-trimethylammonium chloride, 50% in methanol, Thermo Scientific Chemicals
Catalog numberH66414.22
Price (USD)/ Each
72.65
Online exclusive
80.00 
Save 7.35 (9%)
-
Add to cart
Chemical Identifiers
CAS35141-36-7
IUPAC Nametrimethyl[3-(trimethoxysilyl)propyl]azanium chloride
Molecular FormulaC9H24ClNO3Si
InChI KeyFYZFRYWTMMVDLR-UHFFFAOYSA-M
SMILES[Cl-].CO[Si](CCC[N+](C)(C)C)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Color (Gardner Scale)Max 4 Gardner
Assay (Non-aqueous acid-base Titration)48.0-52.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only