2-Chloro-6-(trichloromethyl)pyridine, 98%, Thermo Scientific Chemicals
2-Chloro-6-(trichloromethyl)pyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-6-(trichloromethyl)pyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66418.03
also known as H66418-03
Price (USD)/ Each
37.10
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
37.10
Add to cart
2-Chloro-6-(trichloromethyl)pyridine, 98%, Thermo Scientific Chemicals
Catalog numberH66418.03
Price (USD)/ Each
37.10
-
Add to cart
Chemical Identifiers
CAS1929-82-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale yellow
FormPowder or crystalline powder or solid
Assay (GC)≥97.5%
Melting Point60-67°C
2-Chloro-6-(trichloromethyl)pyridine is

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-6-(trichloromethyl)pyridine is

Solubility
Soluble in 10 mg/ml ethanol.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. L A Robertson; R Cornelisse; R Zeng; J G Kuenen. The effect of thiosulfate and other inhibitors of autotrophic nitrification on heterotrophic nitrifiers. Antonie Van Leeuwenhoek.1989, 56 (4), 301-309.
  2. Iu A Buniatian; M S Petrosian. [Determination of nitrapyrin and its metabolite 6-chloropicolinic acid in the environment]. Hygiene and Sanitation.1983, (4), 52-53.