2-Chloro-6-(trichloromethyl)pyridine is
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloro-6-(trichloromethyl)pyridine is
Solubility
Soluble in 10 mg/ml ethanol.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only
L A Robertson; R Cornelisse; R Zeng; J G Kuenen. The effect of thiosulfate and other inhibitors of autotrophic nitrification on heterotrophic nitrifiers. Antonie Van Leeuwenhoek.1989, 56 (4), 301-309.
Iu A Buniatian; M S Petrosian. [Determination of nitrapyrin and its metabolite 6-chloropicolinic acid in the environment]. Hygiene and Sanitation.1983, (4), 52-53.