2-Amino-2'-chloro-5-nitrobenzophenone, 98%, Thermo Scientific Chemicals
2-Amino-2'-chloro-5-nitrobenzophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-2'-chloro-5-nitrobenzophenone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number H66440.22
also known as H66440-22
Price (USD)/ Each
392.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
392.00
Add to cart
2-Amino-2'-chloro-5-nitrobenzophenone, 98%, Thermo Scientific Chemicals
Catalog numberH66440.22
Price (USD)/ Each
392.00
-
Add to cart
Chemical Identifiers
CAS2011-66-7
IUPAC Name2-(2-chlorobenzoyl)-4-nitroaniline
Molecular FormulaC13H9ClN2O3
InChI KeyGRDGBWVSVMLKBV-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1C(=O)C1=CC=CC=C1Cl)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only