2-[3,4-(Methylenedioxy)phenyl]ethylamine, 95%, Thermo Scientific Chemicals
2-[3,4-(Methylenedioxy)phenyl]ethylamine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-[3,4-(Methylenedioxy)phenyl]ethylamine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66445.03
also known as H66445-03
Price (USD)/ Each
41.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
49.40 
Save 7.75 (16%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
41.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
49.40 
Save 7.75 (16%)
Add to cart
2-[3,4-(Methylenedioxy)phenyl]ethylamine, 95%, Thermo Scientific Chemicals
Catalog numberH66445.03
Price (USD)/ Each
41.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
49.40 
Save 7.75 (16%)
-
Add to cart
Chemical Identifiers
CAS1484-85-1
IUPAC Namehydrogen 2-(2H-1,3-benzodioxol-5-yl)ethan-1-amine chloride
Molecular FormulaC9H12ClNO2
InChI KeyNDYXFQODWGEGNU-UHFFFAOYSA-N
SMILES[H+].[Cl-].NCCC1=CC=C2OCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow
Assay from Supplier's CofA≥94.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only