Indole-2-carbonitrile, 97%, Thermo Scientific Chemicals
Indole-2-carbonitrile, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Indole-2-carbonitrile, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H66451.06
also known as H66451-06
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS36193-65-4
IUPAC Name1H-indole-2-carbonitrile
Molecular FormulaC9H6N2
InChI KeyCBTITARLOCZPDU-UHFFFAOYSA-N
SMILESN#CC1=CC2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
Assay (HPLC)≥96.0%
Proton NMRConforms
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only