5-Acetamidopyridine-3-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals
5-Acetamidopyridine-3-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Acetamidopyridine-3-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
250 mg
Catalog number H66479.MD
also known as H66479-MD
Price (USD)/ Each
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS1201645-46-6
IUPAC NameN-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl]acetamide
Molecular FormulaC13H19BN2O3
InChI KeyADPVUZOCOXRLMP-UHFFFAOYSA-N
SMILESCC(=O)NC1=CN=CC(=C1)B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMR?98%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only