Thermo Scientific Chemicals

Ethyl 6-chloroindole-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: H66483.03
1 g, Each
Thermo Scientific Chemicals

Ethyl 6-chloroindole-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: H66483.03
1 g, Each
Quantity
Catalog number: H66483.03
also known as H66483-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27034-51-1
IUPAC Name
ethyl 6-chloro-1H-indole-2-carboxylate
Molecular Formula
C11H10ClNO2
InChI Key
FSMZLIBWSAMADK-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC2=CC=C(Cl)C=C2N1
Form
Fused solid
Proton NMR
≥96.0% - Conforms
Appearance (Color)
Brown
Melting Point
168.0-173.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text