Thermo Scientific Chemicals

Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97+%, Thermo Scientific Chemicals

Catalog number: H66485.03
1 g, Each
Thermo Scientific Chemicals

Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97+%, Thermo Scientific Chemicals

Catalog number: H66485.03
1 g, Each
Quantity
Catalog number: H66485.03
also known as H66485-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
510758-28-8
IUPAC Name
tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine
Molecular Formula
C30H30N10
InChI Key
WKGZJBVXZWCZQC-UHFFFAOYSA-N
SMILES
C(N(CC1=CN(CC2=CC=CC=C2)N=N1)CC1=CN(CC2=CC=CC=C2)N=N1)C1=CN(CC2=CC=CC=C2)N=N1
Appearance (Color)
White
Assay (unspecified)
≥97.0%
Form
Powder or crystals or crystalline powder
Melting Point
139-146°C

Description

Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine stabilizes Cu(I) towards disproportionation and oxidation thus enhancing its catalytic effect in the azide-acetylene cycloaddition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine stabilizes Cu(I) towards disproportionation and oxidation thus enhancing its catalytic effect in the azide-acetylene cycloaddition.

Solubility
Soluble in DMSO, DMF.

Notes
Store at -20°C. Protect from heat. Incompatible with heat and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text