Chemical Identifiers
CAS170643-02-4
IUPAC Name(2R,3R)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Molecular FormulaC23H27NO5
InChI KeyLZOLWEQBVPVDPR-JLTOFOAXSA-N
SMILESC[C@@H](OC(C)(C)C)[C@@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
View more