Brooker's merocyanine dye is used as solvent polarity indicators. It is used as pH sensors and transition metal cation indicators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Brooker′s merocyanine dye is used as solvent polarity indicators. It is used as pH sensors and transition metal cation indicators.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only
Viviane Cavalli; Domingas C. da Silva; Clodoaldo Machado; Vanderlei G. Machado; Valdir Soldi. The Fluorosolvatochromism of Brooker’s Merocyanine in Pure and in Mixed Solvents. Journal of Fluorescence.2006, 16 77-86.
John O. Morley; Richard M. Morley; Robert Docherty and Michael H. Charlton. Fundamental Studies on Brooker’s Merocyanine. J. Am. Chem. Soc.1997, 119 (42), 10192-10202.