Chemical Identifiers
CAS86060-83-5
IUPAC Name4-(benzyloxy)-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxobutanoic acid
Molecular FormulaC26H23NO6
InChI KeyCBZSVHFNEMONDZ-UHFFFAOYNA-N
SMILESOC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OCC1=CC=CC=C1
View more