N-Boc-L-valine N-succinimidyl ester, 97%, Thermo Scientific Chemicals
N-Boc-L-valine N-succinimidyl ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-L-valine N-succinimidyl ester, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H66559.14
also known as H66559-14
Price (USD)/ Each
63.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
63.00
Add to cart
N-Boc-L-valine N-succinimidyl ester, 97%, Thermo Scientific Chemicals
Catalog numberH66559.14
Price (USD)/ Each
63.00
-
Add to cart
Specifications
Chemical Name or MaterialN-Boc-L-valine N-succinimidyl ester
CAS3392-12-9
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
ColorWhite
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only