N-Boc-L-methionine N-succinimidyl ester, 97%, Thermo Scientific Chemicals
N-Boc-L-methionine N-succinimidyl ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-L-methionine N-succinimidyl ester, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H66563.14
also known as H66563-14
Price (USD)/ Each
241.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
241.00
Add to cart
N-Boc-L-methionine N-succinimidyl ester, 97%, Thermo Scientific Chemicals
Catalog numberH66563.14
Price (USD)/ Each
241.00
-
Add to cart
Specifications
Chemical Name or MaterialN-Boc-L-methionine N-succinimidyl ester
Melting Point120°C to 128°C
CAS3845-64-5
Storage Note 1-30° to -10°C
Recommended StorageStore at -20°C; Store under Nitrogen
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only