Thermo Scientific Chemicals

N-Fmoc-N-methyl-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog number: H66565.06
5 g, Each
Thermo Scientific Chemicals

N-Fmoc-N-methyl-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog number: H66565.06
5 g, Each
Quantity
Catalog number: H66565.06
also known as H66565-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
77128-73-5
IUPAC Name
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)-3-phenylpropanoic acid
Molecular Formula
C25H23NO4
InChI Key
GBROUWPNYVBLFO-UHFFFAOYNA-N
SMILES
CN(C(CC1=CC=CC=C1)C(O)=O)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Assay (HPLC)
>94.0%

Description

N-Fmoc-N-methyl-L-phenylalanine is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-N-methyl-L-phenylalanine is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in DMF. (0.3gram in 2ml)

Notes
Protect from heat. Incompatible with heat and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text