4'-Benzyloxy-2-bromopropiophenone, 97%, Thermo Scientific Chemicals
4'-Benzyloxy-2-bromopropiophenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4'-Benzyloxy-2-bromopropiophenone, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Catalog number H66578.14
also known as H66578-14
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS35081-45-9
IUPAC Name1-[4-(benzyloxy)phenyl]-2-bromopropan-1-one
Molecular FormulaC16H15BrO2
InChI KeyLSQLSCUJBXFBRB-UHFFFAOYNA-N
SMILESCC(Br)C(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
4'-Benzyloxy-2-bromopropiophenone is an intermediate in the preparation of Bazedoxifene acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Benzyloxy-2-bromopropiophenone is an intermediate in the preparation of Bazedoxifene acetate.

Solubility
Soluble in water (2.342g/L at 25°C ).

Notes
Store in freezer -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. C.L. Viswanathan; A.S. Chaudhari. Synthesis and evaluation of uterine relaxant activity for a novel series of substituted p-hydroxyphenylethanolamines. Bioorganic & Medicinal Chemistry. 2006, 14 (19), 6581-6585.
  2. Romero L.S et. al. The effects of 17β-estradiol and a selective estrogen receptor modulator, bazedoxifene, on ovarian carcinogenesis. Gynecologic Oncology. 2012, 124 (1), 134-141.