4-(1-Piperazinyl)indole, 95%, Thermo Scientific Chemicals
4-(1-Piperazinyl)indole, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(1-Piperazinyl)indole, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H66602.MD
also known as H66602-MD
Price (USD)/ Each
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS84807-09-0
IUPAC Name4-(piperazin-1-yl)-1H-indole
Molecular FormulaC12H15N3
InChI KeyYZKSXUIOKWQABW-UHFFFAOYSA-N
SMILESC1CN(CCN1)C1=C2C=CNC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)<0.5%
Appearance (Color)Gray
FormFused solid
Assay from Supplier's CofA≥94.0%
Residue on ignition<0.3%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only