Specifications
Chemical Name or Material(S)-(+)-1-Boc-3-(methylamino)piperidine
CAS912368-73-1
Health Hazard 1H301-H319
Health Hazard 2GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Health Hazard 3P264b-P270-P280i-P301+P310-P305+P351+P338-P330-P501c
View more