D-Valine ethyl ester hydrochloride, 97%, Thermo Scientific Chemicals
D-Valine ethyl ester hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-Valine ethyl ester hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H66619.14
also known as H66619-14
Price (USD)/ Each
473.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
473.00
Add to cart
D-Valine ethyl ester hydrochloride, 97%, Thermo Scientific Chemicals
Catalog numberH66619.14
Price (USD)/ Each
473.00
-
Add to cart
Chemical Identifiers
CAS73913-64-1
IUPAC Nameethyl 2-amino-3-methylbutanoate hydrochloride
Molecular FormulaC7H16ClNO2
InChI KeyPQGVTLQEKCJXKF-UHFFFAOYNA-N
SMILESCl.CCOC(=O)C(N)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only