Thermo Scientific Chemicals

Benzimidazole-6-carbonitrile, 97%, Thermo Scientific Chemicals

Catalog number: H66623.06
5 g, Each
Thermo Scientific Chemicals

Benzimidazole-6-carbonitrile, 97%, Thermo Scientific Chemicals

Catalog number: H66623.06
5 g, Each
Quantity
Catalog number: H66623.06
also known as H66623-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6287-83-8
IUPAC Name
1H-1,3-benzodiazole-6-carbonitrile
Molecular Formula
C8H5N3
InChI Key
NICYTXJGZRYCEQ-UHFFFAOYSA-N
SMILES
N#CC1=CC=C2N=CNC2=C1
Appearance (Color)
Red to orange to brown
Form
Powder
Assay (HPLC)
≥96%
Proton NMR
Conforms to standard

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text