It is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
Maria Keramida; John M. Matsoukas; Dimitris Panagiotopoulos; Kostas Alexopoulos; Elizabeth Matsoukas; John Cladas; Vasilis A. Tzingounis; Emmanuel Cardamakis; Hernâni L. S. Maia, Debanaba Pati; Hamid R. HabibiShow less. Design and synthesis of a gonadotropin-releasing hormone (GnRH) analogue, [Tyr(OMe)5,d-Glu6,Aze9]GnRH: Receptor binding, gonadotropin release and ovulation studies. Letters in Peptide Science. 1996, 3 (4), 257-262.
Hu Huang, Goon Ho Joe; Sung Rok Choi; Su Nam Kim; Yong Tae Kim; Chwang Siek Pak; Joon Hee Hong; and Wonjae Lee. Synthesis of Enantiopure γ-Glutamic Acid Functionalized Peptide Nucleic Acid Monomers. Bull. Korean Chem. Soc. 2010, 31 (7), 2055.