Thermo Scientific Chemicals

Borane-2-methylpyridine complex, 95%, Thermo Scientific Chemicals

Catalog number: H66642.14
25 g, Each
Thermo Scientific Chemicals

Borane-2-methylpyridine complex, 95%, Thermo Scientific Chemicals

Catalog number: H66642.14
25 g, Each
Quantity
Catalog number: H66642.14
also known as H66642-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3999-38-0
IUPAC Name
2-methylpyridine borane
Molecular Formula
C6H10BN
InChI Key
ZMASBEFBIXMNCP-UHFFFAOYSA-N
SMILES
B.CC1=CC=CC=N1
Form
Crystals or powder or crystalline powder
Appearance (Color)
White
Assay from Supplier's CofA
≥94.0%

Description

Used as reactant for N-Benzyl-protection of amino acid derivatives by reductive alkylation, reductive alkoxyamination, as reducing agent for the labeling of oligosaccharides by reductive amination, synthesis of alkoxyamine derivatives, via reduction of oxime ethers, reductive amination reactions of C1-C10 aldehyde 2,4-dinitrophenylhydrazones, synthesis of various trifluoromethylated amino compounds and an alternative reagent for reductive aminations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as reactant for N-Benzyl-protection of amino acid derivatives by reductive alkylation, reductive alkoxyamination, as reducing agent for the labeling of oligosaccharides by reductive amination, synthesis of alkoxyamine derivatives, via reduction of oxime ethers, reductive amination reactions of C1-C10 aldehyde 2,4-dinitrophenylhydrazones, synthesis of various trifluoromethylated amino compounds and an alternative reagent for reductive aminations.

Solubility
Slightly sol. in water, cyclohexane, very sol. in methanol, THF, diglyme, and benzene

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text