N-Acetyl-L-ornithine, 95%, Thermo Scientific Chemicals
N-Acetyl-L-ornithine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Acetyl-L-ornithine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
1 g
Catalog number H66648.06
also known as H66648-06
Price (USD)/ Each
358.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
358.00
Add to cart
N-Acetyl-L-ornithine, 95%, Thermo Scientific Chemicals
Catalog numberH66648.06
Price (USD)/ Each
358.00
-
Add to cart
Chemical Identifiers
CAS6205-08-9
IUPAC Name5-amino-2-acetamidopentanoic acid
Molecular FormulaC7H14N2O3
InChI KeyJRLGPAXAGHMNOL-UHFFFAOYNA-N
SMILESCC(=O)NC(CCCN)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay from Supplier's CofA≥94.0%
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only