Thermo Scientific Chemicals

N-Boc-N-methylethanolamine, 95%, Thermo Scientific Chemicals

Catalog number: H66660.14
25 g, Each
Thermo Scientific Chemicals

N-Boc-N-methylethanolamine, 95%, Thermo Scientific Chemicals

Catalog number: H66660.14
25 g, Each
Quantity
Catalog number: H66660.14
also known as H66660-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57561-39-4
IUPAC Name
tert-butyl N-(2-hydroxyethyl)-N-methylcarbamate
Molecular Formula
C8H17NO3
InChI Key
RFDSJHHLGFFVHD-UHFFFAOYSA-N
SMILES
CN(CCO)C(=O)OC(C)(C)C
Assay (HPLC)
>94.0%

Description

N-Boc-N-methylethanolamine is an substituent in the synthesis of anticancer agent, an orally bioavailable small molecule inhibitor of signal transducer and activator of transcription.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-N-methylethanolamine is an substituent in the synthesis of anticancer agent, an orally bioavailable small molecule inhibitor of signal transducer and activator of transcription.

Solubility
Soluble in diochloromethane. Slightly soluble in water.

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text