Thermo Scientific Chemicals

ITIC-F, Thermo Scientific Chemicals

Catalog number: H66664.MC
100 mg, Each
Thermo Scientific Chemicals

ITIC-F, Thermo Scientific Chemicals

Catalog number: H66664.MC
100 mg, Each
Quantity
Catalog number: H66664.MC
also known as H66664-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2097998-59-7
Form
Flake or threadlike
Bandgap energy
1.55 ? 0.02 eV
Appearance (Color)
Black

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text