1-Cyclopropyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
1-Cyclopropyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Cyclopropyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H66672.MD
also known as H66672-MD
Price (USD)/ Each
99.80
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
99.80
Add to cart
1-Cyclopropyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Catalog numberH66672.MD
Price (USD)/ Each
99.80
-
Add to cart
Chemical Identifiers
CAS1151802-22-0
IUPAC Name1-cyclopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Molecular FormulaC12H19BN2O2
InChI KeyNLWYVKHISUTBMY-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CN(N=C1)C1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
Assay (unspecified)>96.0%
FormSolid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only