6-Bromo-[1,2,4]triazolo[1,5-a]pyridine, 96%, Thermo Scientific Chemicals
6-Bromo-[1,2,4]triazolo[1,5-a]pyridine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Bromo-[1,2,4]triazolo[1,5-a]pyridine, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H66700.MD
also known as H66700-MD
Price (USD)/ Each
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS356560-80-0
IUPAC Name6-bromo-[1,2,4]triazolo[1,5-a]pyridine
Molecular FormulaC6H4BrN3
InChI KeyCXRXKDSDRWLKTK-UHFFFAOYSA-N
SMILESBrC1=CN2N=CN=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)99.80%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only