PDCBT, Thermo Scientific Chemicals
PDCBT, Thermo Scientific Chemicals
Thermo Scientific Chemicals

PDCBT, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
100 mg
500 mg
Catalog number H66713.MD
also known as H66713-MD
Price (USD)/ Each
1,116.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
1,116.00
Add to cart
PDCBT, Thermo Scientific Chemicals
Catalog numberH66713.MD
Price (USD)/ Each
1,116.00
-
Add to cart
Chemical Identifiers
CAS1609536-17-5
SpecificationsSpecification SheetSpecification Sheet
Bandgap energy1.89 ? 0.02 eV
Appearance (Color)Black
FormFlake or threadlike
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only