6-Bromoindole-3-carboxaldehyde, 95%, Thermo Scientific Chemicals
6-Bromoindole-3-carboxaldehyde, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Bromoindole-3-carboxaldehyde, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66756.03
also known as H66756-03
Price (USD)/ Each
100.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
100.00
Add to cart
6-Bromoindole-3-carboxaldehyde, 95%, Thermo Scientific Chemicals
Catalog numberH66756.03
Price (USD)/ Each
100.00
-
Add to cart
Chemical Identifiers
CAS17826-04-9
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Water: ≤0.5%
Melting Point202-206?C
Assay from Supplier's CofA≥94.0%
Appearance (Color)Pale yellow
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only