4-Amino-2-(chloromethyl)pyrimidine, 96%, Thermo Scientific Chemicals
4-Amino-2-(chloromethyl)pyrimidine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Amino-2-(chloromethyl)pyrimidine, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H66760.MD
also known as H66760-MD
Price (USD)/ Each
227.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
227.00
Add to cart
4-Amino-2-(chloromethyl)pyrimidine, 96%, Thermo Scientific Chemicals
Catalog numberH66760.MD
Price (USD)/ Each
227.00
-
Add to cart
Specifications
Chemical Name or Material4-Amino-2-(chloromethyl)pyrimidine
CAS79651-35-7
Health Hazard 1H302-H315-H319-H335
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
ColorColorless to Yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only