Ethyl 5-bromoindole-2-carboxylate, 97%, Thermo Scientific Chemicals
Ethyl 5-bromoindole-2-carboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 5-bromoindole-2-carboxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66797.03
also known as H66797-03
Price (USD)/ Each
32.90
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
32.90
Add to cart
Ethyl 5-bromoindole-2-carboxylate, 97%, Thermo Scientific Chemicals
Catalog numberH66797.03
Price (USD)/ Each
32.90
-
Add to cart
Chemical Identifiers
CAS16732-70-0
IUPAC Nameethyl 5-bromo-1H-indole-2-carboxylate
Molecular FormulaC11H10BrNO2
InChI KeyLWRLKENDQISGEU-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC2=CC(Br)=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
FormFused solid
Melting Point163-167?C
Proton NMRConforms
Assay from Supplier's CofA≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only