Thermo Scientific Chemicals

2-Amino-5-methoxypyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: H66799.MD
250 mg, Each
Thermo Scientific Chemicals

2-Amino-5-methoxypyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: H66799.MD
250 mg, Each
Quantity
Catalog number: H66799.MD
also known as H66799-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13418-77-4
IUPAC Name
5-methoxypyrimidin-2-amine
Molecular Formula
C5H7N3O
InChI Key
KAHHAPNRIQLSFT-UHFFFAOYSA-N
SMILES
COC1=CN=C(N)N=C1
Appearance (Color)
Off-white to yellow
Assay from Suppliers CofA
≥96.0%
Form
Solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text