Specifications
Chemical Name or Material5-Methyl-1-indanone
CAS4593-38-8
Health Hazard 1H302-H319
Health Hazard 2GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Health Hazard 3P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
View more