N-Boc-iminodiacetic acid, 98%, Thermo Scientific Chemicals
N-Boc-iminodiacetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-iminodiacetic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H66868.06
also known as H66868-06
Price (USD)/ Each
69.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
69.90
Add to cart
N-Boc-iminodiacetic acid, 98%, Thermo Scientific Chemicals
Catalog numberH66868.06
Price (USD)/ Each
69.90
-
Add to cart
Chemical Identifiers
CAS56074-20-5
IUPAC Name2-{[(tert-butoxy)carbonyl](carboxymethyl)amino}acetic acid
Molecular FormulaC9H15NO6
InChI KeyQPGIJQUTGABQLQ-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N(CC(O)=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤1.0%
FormPowder
Loss on Drying≤1.0%
Solution TestClear solution (0.3 g in 2mL MeOH)
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only