Thermo Scientific Chemicals

N-Boc-iminodiacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: H66868.14
25 g, Each
Thermo Scientific Chemicals

N-Boc-iminodiacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: H66868.14
25 g, Each
Quantity
Catalog number: H66868.14
also known as H66868-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56074-20-5
IUPAC Name
2-{[(tert-butoxy)carbonyl](carboxymethyl)amino}acetic acid
Molecular Formula
C9H15NO6
InChI Key
QPGIJQUTGABQLQ-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)N(CC(O)=O)CC(O)=O
Water Content (Karl Fischer Titration)
≤1.0%
Form
Powder
Loss on Drying
≤1.0%
Solution Test
Clear solution (0.3 g in 2mL MeOH)
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text