Thermo Scientific Chemicals

2,5-Dimethylindole, 97%, Thermo Scientific Chemicals

Catalog number: H66874.03
1 g, Each
Thermo Scientific Chemicals

2,5-Dimethylindole, 97%, Thermo Scientific Chemicals

Catalog number: H66874.03
1 g, Each
Quantity
Catalog number: H66874.03
also known as H66874-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1196-79-8
IUPAC Name
2,5-dimethyl-1H-indole
Molecular Formula
C10H11N
InChI Key
ZFLFWZRPMDXJCW-UHFFFAOYSA-N
SMILES
CC1=CC2=CC(C)=CC=C2N1
Appearance (Color)
Pale brown to dark brown
Assay (GC)
≥96.0%
Proton NMR
Conforms
Form
Fused solid
Melting Point
112-113?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text