Thermo Scientific Chemicals

3-Amino-6-bromopyrazine-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: H66908.06
5 g, Each
Thermo Scientific Chemicals

3-Amino-6-bromopyrazine-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: H66908.06
5 g, Each
Quantity
Catalog number: H66908.06
also known as H66908-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
486424-37-7
IUPAC Name
3-amino-6-bromopyrazine-2-carboxylic acid
Molecular Formula
C5H4BrN3O2
InChI Key
MTNAQEKMSVDTAQ-UHFFFAOYSA-N
SMILES
NC1=NC=C(Br)N=C1C(O)=O
Assay (HPLC)
>97.5%

Description

3-Amino-6-bromopyrazine-2-carboxylic acid used to prepare 3-bromo-5-phenylpyrazine-2-amine. It serves as an intermediate in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-6-bromopyrazine-2-carboxylic acid used to prepare 3-bromo-5-phenylpyrazine-2-amine. It serves as an intermediate in organic synthesis and pharmaceuticals.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text