N-Fmoc-L-glycylglycine, 97%, Thermo Scientific Chemicals
N-Fmoc-L-glycylglycine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Fmoc-L-glycylglycine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66923.03
also known as H66923-03
Price (USD)/ Each
34.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
40.10 
Save 5.45 (14%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
34.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
40.10 
Save 5.45 (14%)
Add to cart
N-Fmoc-L-glycylglycine, 97%, Thermo Scientific Chemicals
Catalog numberH66923.03
Price (USD)/ Each
34.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
40.10 
Save 5.45 (14%)
-
Add to cart
Chemical Identifiers
CAS35665-38-4
IUPAC Name2-[2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)acetamido]acetic acid
Molecular FormulaC19H18N2O5
InChI KeyFBKUOPULLUJMOC-UHFFFAOYSA-N
SMILESOC(=O)CNC(=O)CNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only