Tetrahydrofuran-3-one, 95%, Thermo Scientific Chemicals
Tetrahydrofuran-3-one, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrahydrofuran-3-one, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H66940.06
also known as H66940-06
Price (USD)/ Each
167.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
167.00
Add to cart
Tetrahydrofuran-3-one, 95%, Thermo Scientific Chemicals
Catalog numberH66940.06
Price (USD)/ Each
167.00
-
Add to cart
Chemical Identifiers
CAS22929-52-8
IUPAC Nameoxolan-3-one
Molecular FormulaC4H6O2
InChI KeyJLPJFSCQKHRSQR-UHFFFAOYSA-N
SMILESO=C1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, brown to yellow
Assay from Supplier's CofA≥94.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only