N-Fmoc-L-valine N-succinimidyl ester is useful for solid phase peptide synthesis techniques.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Fmoc-L-valine N-succinimidyl ester is useful for solid phase peptide synthesis techniques.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
Patrick J. Burke; Brian E. Toki; David W. Meyer; Jamie B. Miyamoto; Kim M. Kissler; Martha Anderson; Peter D. Senter; Scott C. Jeffrey. Novel immunoconjugates comprised of streptonigrin and 17-amino-geldanamycin attached via a dipeptide-p-aminobenzyl-amine linker system. Bioorganic & Medicinal Chemistry Letters, 2009, 19(10), 2650-2653.
Liang Liang; Song-Wen Lin; Wenbing Dai; Jing-Kai Lu; Ting-Yuan Yang; Yu Xiang; Yang Zhang; Run-Tao Li; Qiang Zhang. Novel cathepsin B-sensitive paclitaxel conjugate: Higher water solubility, better efficacy and lower toxicity. Journal of Controlled Release, 2012, (3), 618-629.