5-Amino-2-bromo-4-methylpyridine, 98%, Thermo Scientific Chemicals
5-Amino-2-bromo-4-methylpyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Amino-2-bromo-4-methylpyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H66957.06
also known as H66957-06
Price (USD)/ Each
265.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
265.00
Add to cart
5-Amino-2-bromo-4-methylpyridine, 98%, Thermo Scientific Chemicals
Catalog numberH66957.06
Price (USD)/ Each
265.00
-
Add to cart
Chemical Identifiers
CAS156118-16-0
IUPAC Name6-bromo-4-methylpyridin-3-amine
Molecular FormulaC6H7BrN2
InChI KeyMVDBPMJQCXZKRB-UHFFFAOYSA-N
SMILESCC1=CC(Br)=NC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to pale brown
Assay from Suppliers CofA≥97.5%
FormSolid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only