4-Bromoindole-3-carboxaldehyde, 97%, Thermo Scientific Chemicals
4-Bromoindole-3-carboxaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromoindole-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66964.03
also known as H66964-03
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS98600-34-1
IUPAC Name4-bromo-1H-indole-3-carbaldehyde
Molecular FormulaC9H6BrNO
InChI KeyIPAFHZDRYAWZOB-UHFFFAOYSA-N
SMILESBrC1=C2C(NC=C2C=O)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMR≥96.0% - Conforms
Appearance (Color)Gray
FormFused solid
Assay (HPLC)≥96.0%
Melting Point175-177?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only