Invitrogen™

SureCast™ Gel Handcast Bundle A - Hardware and Reagents

Catalog number: HC1000SR
Have Questions?
Invitrogen™

SureCast™ Gel Handcast Bundle A - Hardware and Reagents

Catalog number: HC1000SR
Have Questions?
Catalog Number
HC1000SR
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
HC1000SR1 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Pour your own polyacrylamide gels easily and confidently with a SureCast™ Gel Handcast Bundle A. The complete product includes the SureCast™ Handcast System and SureCast™ Handcast Reagents. For a bundle that does not include the SureCast Handcast Reagents, please see SureCast™ Gel Handcast Bundle B. The system features durable glass plates that are securely sealed using a single-motion, load-and-lock mechanism. The unique tilt feature of the SureCast™ Handcast Station minimizes spillage and enhances loadability of casting solutions during handcasting. SureCast gels are designed for use in the Mini Gel Tank.

Features of the SureCast Handcast System include:
• Innovate, leak-free design—minimizes the number of failed gels, resulting in less wasted time
• Superior glass plate durability
• Simple assembly of casting components—just drop the glass plates in, then lock the cam handle
• Unique tilt feature for easy loading of acrylamide solutions

The SureCast Handcast System can be used to create polyacrylamide gels using SureCast Handcast Reagents as well as any other popular polyacrylamide gel casting reagents.

The SureCast Handcast Bundle A includes all the hardware and reagents you need to get started:
2 SureCast Handcast Stations
2 SureCast™ Glass Plate Sets (2 front and 2 back)
6 SureCast™ Gel Combs (2 each: 10-, 12-, 15-well)
10 SureCast™ Gel Spacers
1 SureCast™ Stacking Buffer (1 L), 2-pack
1 SureCast™ Resolving Buffer (1 L), 2-pack
1 SureCast™ Acrylamide Solution (40%) (450 mL)
1 SureCast™ APS (25 g)
1 SureCast™ TEMED (30 mL)

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gel Size
Mini
Product Line
SureCast
Type
Gel Handcast Bundle A
Shipping Condition
Room Temperature

Contents & Storage

• 2 SureCast Gel Handcast Stations
• 2 sets SureCast Glass Plates
• 10 SureCast Gel Spacers
• 1 SureCast 10-well Multi-Use Tool
• 1 SureCast 12-well Multi-Use Tool
• 1 SureCast 15-well Multi-Use Tool
• 1 set SureCast Gel Combs (10-, 12-, 15-well)
• 2 x 500 mL SureCast Stacking Buffer packs.

Store at room temperature.
• 5 x 500 mL SureCast Stacking Buffer packs.

Store at room temperature.
• 2 x 500 mL SureCast Resolving Buffer packs.

Store at room temperature.
• 5 x 500 mL SureCast Resolving Buffer packs.

Store at room temperature.
• 25 g SureCast Ammonium Persulfate.

Store at room temperature.
• 450 mL SureCast Acrylamade Solution.

Store at room temperature.
• 30 mL SureCast TEMED.

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text