SureCast™ Glass Plates are used to cast 1.0 mm SureCast™ polyacrylamide gels using the SureCast™ Handcast Station, offering superior durabilityRead more
Have Questions?
Catalog number HC1001
Price (USD)/ 2 sets
133.65
Online exclusive
137.00
Save 3.35 (2%)
-
Add to cart
Price (USD)/ 2 sets
133.65
Online exclusive
137.00
Save 3.35 (2%)
Add to cart
SureCast™ Glass Plates
Catalog numberHC1001
Price (USD)/ 2 sets
133.65
Online exclusive
137.00
Save 3.35 (2%)
-
Add to cart
SureCast™ Glass Plates are used to cast 1.0 mm SureCast™ polyacrylamide gels using the SureCast™ Handcast Station, offering superior durability and resistance to damage. These gels are designed for use in the Mini Gel Tank. SureCast™ Gel Spacers are not included and are sold separately.
Includes: 2 front glass plates 2 back glass plates